Assay Design
Gene Transcripts
- Gene Symbol
- pro-3
- Entrez Gene ID
-
180256
- Gene Name
- Protein SDA1 homolog
- Gene Aliases
-
CELE_Y39B6A.14
- Chromosome Location
-
Chr.V: 19101337 - 19118412 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171594.5 |
NP_741684.2 |
7 - 8 |
2109 |
59 |
View Full Product Details