Assay Design



Gene Transcripts

Gene Symbol
bed-1
Entrez Gene ID
180257
Gene Name
protein-coding
Gene Aliases
CELE_Y39B6A.12
Chromosome Location
Chr.V: 19119527 - 19124922 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029056.5 NP_001024227.1 2 - 3 376 130
NM_001029057.2 NP_001024228.1 2 - 3 369 130

View Full Product Details