Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270046.2 | NP_001256975.1 | 8 - 9 | 1692 | 140 |
NM_001374936.2 | NP_001362085.1 | 21 - 22 | 3791 | 140 | |
NM_001380893.1 | NP_001367765.1 | 18 - 19 | 3572 | 140 |