Assay Design
Gene Transcripts
- Gene Symbol
- aex-3
- Entrez Gene ID
-
180420
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C02H7.3
- Chromosome Location
-
Chr.X: 751145 - 759734 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171618.5 |
NP_741710.1 |
18 - 19 |
4132 |
120 |
GenBank mRNA |
U93842.1 |
- |
18 - 19 |
4154 |
120 |
View Full Product Details