Assay Design



Gene Transcripts

Gene Symbol
aex-3
Entrez Gene ID
180420
Gene Name
protein-coding
Gene Aliases
CELE_C02H7.3
Chromosome Location
Chr.X: 751145 - 759734 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029171.7 NP_001024342.1 7 - 8 1312 67
NM_171618.5 NP_741710.1 7 - 8 1310 67
GenBank mRNA U93842.1 - 7 - 8 1332 67

View Full Product Details