Assay Design



Gene Transcripts

Gene Symbol
klp-4
Entrez Gene ID
180608
Gene Name
protein-coding
Gene Aliases
CELE_F56E3.3
Chromosome Location
Chr.X: 3171420 - 3184780 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373256.3 NP_001359863.1 17 - 17 4068 105
NM_001373257.3 NP_001359864.1 17 - 17 4023 105

View Full Product Details