Assay Design



Gene Transcripts

Gene Symbol
daf-9
Entrez Gene ID
180889
Gene Name
protein-coding
Gene Aliases
CELE_T13C5.1
Chromosome Location
Chr.X: 6196927 - 6200823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029732.3 NP_001024903.1 2 - 3 403 152
NM_001375044.2 NP_001362132.1 1 - 2 340 152
NM_171699.6 NP_741807.2 3 - 4 509 152
GenBank mRNA AF407572.1 - 2 - 3 403 152

View Full Product Details