Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001381028.1 | NP_001367692.1 | 9 - 10 | 1132 | 84 |
NM_001381029.1 | NP_001368577.1 | 9 - 10 | 1056 | 84 | |
NM_001381030.1 | NP_001367691.1 | 9 - 10 | 1132 | 84 | |
NM_001381031.2 | NP_001367690.1 | 9 - 10 | 1132 | 84 | |
NM_171733.5 | NP_741848.1 | 11 - 12 | 1218 | 84 | |
GenBank mRNA | AF368920.1 | - | 9 - 10 | 1081 | 84 |
AY313898.1 | - | 9 - 10 | 1132 | 84 | |
AY313899.1 | - | 9 - 10 | 1132 | 84 | |
AY313900.1 | - | 9 - 10 | 1132 | 84 | |
AY322480.1 | - | 9 - 10 | 1132 | 84 |