Assay Design



Gene Transcripts

Gene Symbol
grd-1
Entrez Gene ID
181300
Gene Name
GRounDhog (hedgehog-like family)
Gene Aliases
CELE_R08B4.1
Chromosome Location
Chr.X: 11109440 - 11115887 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029672.3 NP_001024843.1 3 - 4 600 126

View Full Product Details