Assay Design



Gene Transcripts

Gene Symbol
ifa-1
Entrez Gene ID
181316
Gene Name
protein-coding
Gene Aliases
CELE_F38B2.1
Chromosome Location
Chr.X: 11271184 - 11275498 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083295.4 NP_001076764.1 1 - 2 69 67
NM_001270210.4 NP_001257139.1 2 - 3 142 67
NM_001373503.1 NP_001360640.1 2 - 3 178 67
NM_171777.8 NP_741902.1 2 - 3 155 67
NM_171778.7 NP_741903.1 2 - 3 197 67

View Full Product Details