Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083295.4 | NP_001076764.1 | 5 - 6 | 1309 | 65 |
NM_001270210.4 | NP_001257139.1 | 6 - 7 | 1382 | 65 | |
NM_001373503.1 | NP_001360640.1 | 6 - 7 | 1418 | 65 | |
NM_171777.8 | NP_741902.1 | 6 - 7 | 1395 | 65 | |
NM_171778.7 | NP_741903.1 | 6 - 7 | 1437 | 65 |