Assay Design



Gene Transcripts

Gene Symbol
nhr-71
Entrez Gene ID
181491
Gene Name
protein-coding
Gene Aliases
CELE_K11E4.5
Chromosome Location
Chr.X: 13747315 - 13751797 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029616.6 NP_001024787.1 3 - 4 371 69
NM_001029617.5 NP_001024788.1 3 - 4 248 69

View Full Product Details