Assay Design



Gene Transcripts

Gene Symbol
cdc-26
Entrez Gene ID
172958
Gene Name
protein-coding
Gene Aliases
CELE_B0511.9
Chromosome Location
Chr.I: 10639076 - 10640234 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264516.3 NP_001251445.1 3 - 4 207 66
NM_171844.6 NP_740914.2 2 - 3 262 66

View Full Product Details