Assay Design



Gene Transcripts

Gene Symbol
ife-3
Entrez Gene ID
178536
Gene Name
protein-coding
Gene Aliases
CELE_B0348.6
Chromosome Location
Chr.V: 6584 - 7829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070722.5 NP_503123.1 1 - 2 175 74
NM_070723.7 NP_503124.1 1 - 2 168 74
NM_171920.6 NP_741502.1 1 - 2 174 74

View Full Product Details