Assay Design
Gene Transcripts
- Gene Symbol
- ife-3
- Entrez Gene ID
-
178536
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_B0348.6
- Chromosome Location
-
Chr.V: 6584 - 7829 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171920.6 |
NP_741502.1 |
2 - 3 |
352 |
63 |
View Full Product Details