Assay Design
Gene Transcripts
- Gene Symbol
- sdn-1
- Entrez Gene ID
-
181259
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F57C7.3
- Chromosome Location
-
Chr.X: 10589770 - 10593334 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171972.7 |
NP_741894.1 |
1 - 2 |
210 |
84 |
View Full Product Details