Assay Design
Gene Transcripts
- Gene Symbol
- ztf-1
- Entrez Gene ID
-
176236
- Gene Name
- Zinc finger transcription factor family protein 1
- Gene Aliases
-
CELE_F54F2.5
- Chromosome Location
-
Chr.III: 8787092 - 8792351 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066541.5 |
NP_498942.1 |
12 - 13 |
1588 |
109 |
View Full Product Details