Assay Design



Gene Transcripts

Gene Symbol
set-3
Entrez Gene ID
182356
Gene Name
protein-coding
Gene Aliases
CELE_C07A9.7
Chromosome Location
Chr.III: 9691027 - 9694574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066742.5 NP_499143.1 2 - 3 361 104
NM_181940.6 NP_871669.1 2 - 3 363 104

View Full Product Details