Assay Design
Gene Transcripts
- Gene Symbol
- col-96
- Entrez Gene ID
-
353461
- Gene Name
- Col_cuticle_N domain-containing protein
- Gene Aliases
-
CELE_Y41C4A.19
- Chromosome Location
-
Chr.III: 11705652 - 11706765 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_181982.4 |
NP_871711.1 |
1 - 2 |
204 |
60 |
View Full Product Details