Assay Design



Gene Transcripts

Gene Symbol
ubc-26
Entrez Gene ID
259424
Gene Name
protein-coding
Gene Aliases
CELE_Y110A2AM.3
Chromosome Location
Chr.II: 2894771 - 2900528 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001361850.2 NP_001348718.1 2 - 3 272 103
NM_001383804.1 3 - 4 281 103

View Full Product Details