Assay Design
Gene Transcripts
- Gene Symbol
- btb-2
- Entrez Gene ID
-
174061
- Gene Name
- BTB domain-containing protein
- Gene Aliases
-
CELE_T25D10.5
- Chromosome Location
-
Chr.II: 6401949 - 6403105 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182195.5 |
NP_871995.1 |
3 - 4 |
645 |
71 |
View Full Product Details