Assay Design



Gene Transcripts

Gene Symbol
clec-88
Entrez Gene ID
174056
Gene Name
protein-coding
Gene Aliases
CELE_K10B2.3
Chromosome Location
Chr.II: 6365722 - 6367002 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062882.9 NP_495283.2 1 - 2 178 88
NM_182210.5 NP_872010.1 1 - 2 196 88
GenBank mRNA DQ340628.1 - 1 - 2 141 88

View Full Product Details