Assay Design



Gene Transcripts

Gene Symbol
prdx-2
Entrez Gene ID
266858
Gene Name
protein-coding
Gene Aliases
CELE_F09E5.15
Chromosome Location
Chr.II: 5377704 - 5379633 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313607.3 NP_001300536.1 2 - 3 391 64
NM_001381416.1 NP_001367731.1 1 - 2 385 64
NM_001383831.1 2 - 3 530 64

View Full Product Details