Assay Design
Gene Transcripts
- Gene Symbol
- flp-1
- Entrez Gene ID
-
177737
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F23B2.5
- Chromosome Location
-
Chr.IV: 9144692 - 9146128 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182277.8 |
NP_872077.1 |
3 - 4 |
255 |
92 |
View Full Product Details