Assay Design
Gene Transcripts
- Gene Symbol
- acdh-13
- Entrez Gene ID
-
177654
- Gene Name
- Acyl CoA DeHydrogenase
- Gene Aliases
-
CELE_C28C12.9
- Chromosome Location
-
Chr.IV: 8496484 - 8498823 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069051.6 |
NP_501452.1 |
5 - 6 |
1183 |
106 |
View Full Product Details