Assay Design
Gene Transcripts
- Gene Symbol
- srv-26
- Entrez Gene ID
-
353424
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y73B6BL.40
- Chromosome Location
-
Chr.IV: 6299273 - 6301864 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182313.1 |
NP_872113.1 |
5 - 6 |
640 |
93 |
View Full Product Details