Assay Design



Gene Transcripts

Gene Symbol
twk-43
Entrez Gene ID
3565024
Gene Name
protein-coding
Gene Aliases
CELE_F32H5.7
Chromosome Location
Chr.V: 13346278 - 13356168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276947.3 NP_001263876.1 7 - 8 813 69
NM_001276948.3 NP_001263877.1 7 - 8 786 69
NM_001276949.3 NP_001263878.1 4 - 5 555 69
NM_182337.7 NP_872137.2 8 - 9 893 69

View Full Product Details