Assay Design



Gene Transcripts

Gene Symbol
daf-16
Entrez Gene ID
172981
Gene Name
protein-coding
Gene Aliases
CELE_R13H8.1
Chromosome Location
Chr.I: 10750498 - 10776703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026422.6 NP_001021593.1 2 - 3 806 71
NM_001026425.6 NP_001021596.1 6 - 7 677 71
NM_001026427.7 NP_001021598.2 7 - 8 767 71
NM_001377627.2 NP_001364785.1 5 - 6 746 71
NM_001377628.3 NP_001364784.1 5 - 6 740 71
NM_001381205.1 NP_001367893.1 7 - 8 839 71
NM_001381206.1 NP_001368498.1 7 - 8 761 71
NM_001381207.1 NP_001368499.1 6 - 7 671 71
NM_001381208.1 NP_001368501.1 4 - 5 437 71
NM_001381209.1 NP_001368502.1 4 - 5 431 71
NM_001381210.1 NP_001367892.1 1 - 2 125 71
GenBank mRNA AF020342.1 - 5 - 6 1044 71
AF020343.1 - 5 - 6 1036 71
AF020344.1 - 2 - 3 855 71
AF032112.1 - 5 - 6 1079 71

View Full Product Details