Assay Design



Gene Transcripts

Gene Symbol
daf-16
Entrez Gene ID
172981
Gene Name
protein-coding
Gene Aliases
CELE_R13H8.1
Chromosome Location
Chr.I: 10750498 - 10776703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026425.6 NP_001021596.1 4 - 5 436 82
NM_001026427.7 NP_001021598.2 5 - 6 526 82
NM_001377627.2 NP_001364785.1 3 - 4 505 82
NM_001377628.3 NP_001364784.1 3 - 4 499 82
NM_001381205.1 NP_001367893.1 5 - 6 598 82
NM_001381206.1 NP_001368498.1 5 - 6 520 82
NM_001381207.1 NP_001368499.1 4 - 5 430 82
NM_001381208.1 NP_001368501.1 2 - 3 196 82
NM_001381209.1 NP_001368502.1 2 - 3 190 82
GenBank mRNA AF020342.1 - 3 - 4 803 82
AF020343.1 - 3 - 4 795 82
AF032112.1 - 3 - 4 838 82

View Full Product Details