Assay Design
Gene Transcripts
- Gene Symbol
- lrk-1
- Entrez Gene ID
-
172995
- Gene Name
- Leucine-rich repeat serine/threonine-protein kinase 1
- Gene Aliases
-
CELE_T27C10.6
- Chromosome Location
-
Chr.I: 10879769 - 10894239 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060438.7 |
NP_492839.4 |
18 - 19 |
6860 |
64 |
GenBank mRNA |
AB297384.1 |
- |
18 - 19 |
6846 |
64 |
View Full Product Details