Assay Design



Gene Transcripts

Gene Symbol
srt-61
Entrez Gene ID
188990
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_T27C10.1
Chromosome Location
Chr.I: 10904370 - 10906766 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377631.2 NP_001364776.1 2 - 3 336 157

View Full Product Details