Assay Design
Gene Transcripts
- Gene Symbol
- clec-95
- Entrez Gene ID
-
173010
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_ZK39.2
- Chromosome Location
-
Chr.I: 11148973 - 11150303 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060467.6 |
NP_492868.1 |
3 - 4 |
344 |
55 |
View Full Product Details