Assay Design
Gene Transcripts
- Gene Symbol
- clec-96
- Entrez Gene ID
-
191213
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_ZK39.5
- Chromosome Location
-
Chr.I: 11151224 - 11151975 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060468.3 |
NP_492869.1 |
2 - 3 |
232 |
61 |
View Full Product Details