Assay Design
Gene Transcripts
- Gene Symbol
- clec-99
- Entrez Gene ID
-
173011
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_ZK39.8
- Chromosome Location
-
Chr.I: 11156728 - 11158833 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060471.6 |
NP_492872.1 |
1 - 2 |
99 |
84 |
View Full Product Details