Assay Design



Gene Transcripts

Gene Symbol
cdk-9
Entrez Gene ID
173023
Gene Name
protein-coding
Gene Aliases
CELE_H25P06.2
Chromosome Location
Chr.I: 11323527 - 11333154 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060505.4 NP_492906.2 2 - 3 108 75
NM_060506.5 NP_492907.2 2 - 3 108 75

View Full Product Details