Assay Design



Gene Transcripts

Gene Symbol
spe-9
Entrez Gene ID
173042
Gene Name
protein-coding
Gene Aliases
CELE_C17D12.6
Chromosome Location
Chr.I: 11582743 - 11586069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136278.3 NP_001129750.1 5 - 6 611 63
NM_060554.5 NP_492955.1 12 - 13 1476 63
GenBank mRNA AF049327.1 - 12 - 13 1454 63

View Full Product Details