Assay Design
Gene Transcripts
- Gene Symbol
- spe-9
- Entrez Gene ID
-
173042
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C17D12.6
- Chromosome Location
-
Chr.I: 11582743 - 11586069 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060554.5 |
NP_492955.1 |
5 - 6 |
638 |
79 |
GenBank mRNA |
AF049327.1 |
- |
5 - 6 |
616 |
79 |
View Full Product Details