Assay Design



Gene Transcripts

Gene Symbol
spe-9
Entrez Gene ID
173042
Gene Name
protein-coding
Gene Aliases
CELE_C17D12.6
Chromosome Location
Chr.I: 11582743 - 11586069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136278.3 NP_001129750.1 2 - 3 169 73
NM_060554.5 NP_492955.1 9 - 10 1034 73
GenBank mRNA AF049327.1 - 9 - 10 1012 73

View Full Product Details