Assay Design
Gene Transcripts
- Gene Symbol
- dhhc-2
- Entrez Gene ID
-
173062
- Gene Name
- Palmitoyltransferase
- Gene Aliases
-
CELE_Y47H9C.2
- Chromosome Location
-
Chr.I: 11836460 - 11849258 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060606.6 |
NP_493007.2 |
5 - 6 |
823 |
157 |
View Full Product Details