Assay Design
Gene Transcripts
- Gene Symbol
- oac-35
- Entrez Gene ID
-
186421
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_F56H6.11
- Chromosome Location
-
Chr.I: 12307731 - 12310431 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060710.2 |
NP_493111.1 |
4 - 5 |
597 |
88 |
View Full Product Details