Assay Design
Gene Transcripts
- Gene Symbol
- glct-1
- Entrez Gene ID
-
188329
- Gene Name
- Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase
- Gene Aliases
-
CELE_T09E11.1
- Chromosome Location
-
Chr.I: 12337968 - 12339239 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060720.2 |
NP_493121.2 |
2 - 3 |
180 |
92 |
View Full Product Details