Assay Design



Gene Transcripts

Gene Symbol
gly-15
Entrez Gene ID
173114
Gene Name
protein-coding
Gene Aliases
CELE_C54C8.11
Chromosome Location
Chr.I: 12462176 - 12465816 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276610.2 NP_001263539.1 1 - 2 89 102
NM_060763.4 NP_493164.2 1 - 2 89 102

View Full Product Details