Assay Design
Gene Transcripts
- Gene Symbol
- gly-15
- Entrez Gene ID
-
173114
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C54C8.11
- Chromosome Location
-
Chr.I: 12462176 - 12465816 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060763.4 |
NP_493164.2 |
4 - 5 |
588 |
66 |
View Full Product Details