Assay Design



Gene Transcripts

Gene Symbol
par-6
Entrez Gene ID
173137
Gene Name
protein-coding
Gene Aliases
CELE_T26E3.3
Chromosome Location
Chr.I: 12669380 - 12674449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047222.6 NP_001040687.1 7 - 8 810 69
NM_001047223.5 NP_001040688.1 4 - 5 372 69
GenBank mRNA AF070968.1 - 7 - 8 809 69

View Full Product Details