Assay Design
Gene Transcripts
- Gene Symbol
- ndx-1
- Entrez Gene ID
-
173138
- Gene Name
- Putative nudix hydrolase 1
- Gene Aliases
-
CELE_T26E3.2
- Chromosome Location
-
Chr.I: 12679207 - 12681818 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060808.3 |
NP_493209.1 |
2 - 3 |
192 |
72 |
View Full Product Details