Assay Design
Gene Transcripts
- Gene Symbol
- sra-22
- Entrez Gene ID
-
185059
- Gene Name
- Serpentine receptor class alpha-22
- Gene Aliases
-
CELE_F28C12.7
- Chromosome Location
-
Chr.I: 12701439 - 12702886 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060817.2 |
NP_493218.2 |
2 - 3 |
393 |
153 |
View Full Product Details