Assay Design
Gene Transcripts
- Gene Symbol
- fbxb-66
- Entrez Gene ID
-
189784
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y40B1B.3
- Chromosome Location
-
Chr.I: 13423997 - 13425148 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060961.6 |
NP_493362.1 |
1 - 2 |
97 |
112 |
View Full Product Details