Assay Design
Gene Transcripts
- Gene Symbol
- ndx-8
- Entrez Gene ID
-
190780
- Gene Name
- Peroxisomal coenzyme A diphosphatase ndx-8
- Gene Aliases
-
CELE_Y87G2A.14
- Chromosome Location
-
Chr.I: 13492983 - 13507068 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060971.3 |
NP_493372.1 |
3 - 4 |
311 |
81 |
View Full Product Details