Assay Design
Gene Transcripts
- Gene Symbol
- vps-28
- Entrez Gene ID
-
173229
- Gene Name
- Vacuolar protein sorting-associated protein 28 homolog
- Gene Aliases
-
CELE_Y87G2A.10
- Chromosome Location
-
Chr.I: 13586037 - 13588831 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060981.6 |
NP_493382.1 |
2 - 3 |
365 |
94 |
View Full Product Details