Assay Design
Gene Transcripts
- Gene Symbol
- hsd-1
- Entrez Gene ID
-
189371
- Gene Name
- 3Beta_HSD domain-containing protein
- Gene Aliases
-
CELE_Y6B3B.11
- Chromosome Location
-
Chr.I: 13720125 - 13724453 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061001.1 |
NP_493402.1 |
1 - 2 |
115 |
76 |
View Full Product Details